2,5′-Dibrom-2′-hydroxyacetophenone, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Bromo-1-(5-bromo-2-hydroxyphenyl)ethanone
M-Clarity™ Qualitätsstufe = MQ100
663670-1G
663670-5G
SIAL663670-1GEA
42.7
EUR
SIAL663670-1G
SIAL663670-5G
2,5′-Dibrom-2′-hydroxyacetophenone, Sigma-Aldrich®
2,5′-Dibrom-2′-hydroxyacetophenone
Formel:
C₈H₆Br₂O₂ Molecular Weight: 293,9400 g/mol |
MDL:
MFCD01067749 CAS-Nummer: 67029-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...