4'-Brom-2'-hydroxypropiophenon

Lieferant: BIONET RESEARCH

Synonyme: 1-(4-Bromo-2-hydroxyphenyl)propan-1-one

9T-0062.500 9T-0062.1 9T-0062.5 9T-0062.10
BIOT9T-0062.500EA 0 EUR
BIOT9T-0062.500 BIOT9T-0062.1 BIOT9T-0062.5 BIOT9T-0062.10
4'-Brom-2'-hydroxypropiophenon
4'-Brom-2'-hydroxypropiophenon
Formel: C₉H₉BrO₂
MDL: MFCD03791251
CAS-Nummer: 17764-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR