5'-Brom-2'-hydroxypropiophenon

Lieferant: CHEMPUR

Synonyme: 1-(5-Bromo-2-hydroxyphenyl)propan-1-one

FL12088.POR 012088-25G 012088-5G
CHMPFL12088.POREA 0 EUR
CHMPFL12088.POR FLUO012088-25G FLUO012088-5G
5'-Brom-2'-hydroxypropiophenon
5'-Brom-2'-hydroxypropiophenon
Formel: C₉H₉BrO₂
Molecular Weight: 229,0706 g/mol
MDL: MFCD00218577
CAS-Nummer: 17764-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR