2-Amino-4-chlor-2'-fluorbenzophenon

Lieferant: FLUOROCHEM

003840-250MG 003840-1G
FLUO003840-250MGEA 137 EUR
FLUO003840-250MG FLUO003840-1G
2-Amino-4-chlor-2'-fluorbenzophenon
2-Amino-4-chlor-2'-fluorbenzophenon
Formel: C₁₃H₉ClFNO
MDL: MFCD00236584
CAS-Nummer: 203303-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR