4'-Chlor-2'-fluor-5'-methylacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(4-Chloro-2-fluoro-5-methylphenyl)ethanone
003855-1G
003855-10G
FLUO003855-1GEA
119
EUR
FLUO003855-1G
FLUO003855-10G
4'-Chlor-2'-fluor-5'-methylacetophenon
4'-Chlor-2'-fluor-5'-methylacetophenon
Formel:
C₉H₈ClFO |
MDL:
MFCD00236601 CAS-Nummer: 177211-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...