5'-Ethyl-2'-hydroxy-3'-nitroacetophenon

Lieferant: CHEMPUR

FL12083.POR 012083-5G 012083-1G
CHMPFL12083.POREA 0 EUR
CHMPFL12083.POR FLUO012083-5G FLUO012083-1G
5'-Ethyl-2'-hydroxy-3'-nitroacetophenon
5'-Ethyl-2'-hydroxy-3'-nitroacetophenon
Formel: C₁₀H₁₁NO₄
MDL: MFCD03424515
CAS-Nummer: 71002-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR