2-Fluor-6-(trifluormethyl)phenylacetonitril

Lieferant: FLUOROCHEM

006444-1G 006444-5G
FLUO006444-1GEA 48.8 EUR
FLUO006444-1G FLUO006444-5G
2-Fluor-6-(trifluormethyl)phenylacetonitril
2-Fluor-6-(trifluormethyl)phenylacetonitril
Formel: C₉H₅F₄N
MDL: MFCD00061184
CAS-Nummer: 179946-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR