2-Brom-5-chlor-4-methylanilin

Lieferant: COMBI-BLOCKS

AN-1264-5G AN-1264-1G
COBBAN-1264-5GEA 711 EUR
COBBAN-1264-5G COBBAN-1264-1G
2-Brom-5-chlor-4-methylanilin
2-Brom-5-chlor-4-methylanilin
Formel: C₇H₇BrClN
Molecular Weight: 220,4942 g/mol
MDL: MFCD00806533
CAS-Nummer: 102170-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR