4-Brom-3-chlor-2-methylanilin

Lieferant: COMBI-BLOCKS

AN-1158-100G AN-1158-5G AN-1158-25G
COBBAN-1158-100GEA 884 EUR
COBBAN-1158-100G COBBAN-1158-5G COBBAN-1158-25G
4-Brom-3-chlor-2-methylanilin
4-Brom-3-chlor-2-methylanilin
Formel: C₇H₇BrClN
Molecular Weight: 220,4942 g/mol
MDL: MFCD07369999
CAS-Nummer: 627531-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR