1,1,1,3,3,3-Hexafluorisopropylacrylat
Lieferant: FLUOROCHEM
Synonyme:
1,1,1,3,3,3-Hexafluoropropan-2-yl acrylate
003254-10G
003254-25G
003254-100G
FLUO003254-10GEA
48.8
EUR
FLUO003254-10G
FLUO003254-25G
FLUO003254-100G
1,1,1,3,3,3-Hexafluorisopropylacrylat
1,1,1,3,3,3-Hexafluorisopropylacrylat
Formel:
C₆H₄F₆O₂ Molecular Weight: 222,09 g/mol |
MDL:
MFCD00040104 CAS-Nummer: 2160-89-6 EINECS: 218-479-1 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...