1H,1H,3H-Hexafluorbutylacrylat

Lieferant: FLUOROCHEM

Synonyme: HFBA, 2,2,3,4,4,4-Hexafluorobutyl Acrylate, Acrylic acid 2,2,3,4,4,4-Hexafluorobutyl ester

002399-25G 002399-5G
FLUO002399-25GEA 248 EUR
FLUO002399-25G FLUO002399-5G
1H,1H,3H-Hexafluorbutylacrylat
1H,1H,3H-Hexafluorbutylacrylat
Formel: C₇H₆F₆O₂
Molecular Weight: 236,1118 g/mol
MDL: MFCD00044091
CAS-Nummer: 54052-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR