1,1,1,3,3,3-Hexafluorisopropylmethacrylat
Lieferant: FLUOROCHEM
Synonyme:
Hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-Hexafluoroisopropylmethacrylate
003255-10G
003255-50G
003255-100G
FLUO003255-10GEA
0
EUR
FLUO003255-10G
FLUO003255-50G
FLUO003255-100G
1,1,1,3,3,3-Hexafluorisopropylmethacrylat
1,1,1,3,3,3-Hexafluorisopropylmethacrylat
Formel:
C₇H₆F₆O₂ Molecular Weight: 236.12 g/mol |
MDL:
MFCD00040105 CAS-Nummer: 3063-94-3 EINECS: 221-309-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...