6-Bromanthranilsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-6-bromobenzoic acid

H32106.03 H32106.06
H32106.03EA 55.3 EUR
H32106.03 H32106.06
6-Bromanthranilsäure ≥98%
6-Bromanthranilsäure

Purity: 98% CAS No.: 20776-48-1 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 MDL No.: MFCD03618455

Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03618455
CAS-Nummer: 20776-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR