5-Amino-2-brombenzoesäure

Lieferant: COMBI-BLOCKS

HC-2067-5G HC-2067-25G HC-2067-1G
COBBHC-2067-5GEA 63.9 EUR
COBBHC-2067-5G COBBHC-2067-25G COBBHC-2067-1G
5-Amino-2-brombenzoesäure
5-Amino-2-brombenzoesäure
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
MDL: MFCD07368968
CAS-Nummer: 2840-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR