5-Amino-2-brombenzoesäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
668036-25G
668036-5G
SIAL668036-25GEA
311
EUR
SIAL668036-25G
SIAL668036-5G
5-Amino-2-brombenzoesäure, Sigma-Aldrich®
5-Amino-2-brombenzoesäure
Formel:
C₇H₆BrNO₂ Molecular Weight: 216,03 g/mol |
MDL:
MFCD07368968 CAS-Nummer: 2840-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...