4-Amino-3-brombenzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-3-bromobenzoic Acid

M-Clarity™ Qualitätsstufe = MQ100

663484-10G 663484-1G
SIAL663484-10GEA 159 EUR
SIAL663484-10G SIAL663484-1G
4-Amino-3-brombenzoesäure, Sigma-Aldrich®
4-Amino-3-bromobenzoic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216.0320 g/mol
MDL: MFCD03407439
CAS-Nummer: 6311-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR