2-Brom-4-chlor-6-nitroanilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-4-chloro-6-nitrophenylamine

HC-2904-5G HC-2904-1G
COBBHC-2904-5GEA 33.4 EUR
COBBHC-2904-5G COBBHC-2904-1G
2-Brom-4-chlor-6-nitroanilin
2-Brom-4-chlor-6-nitroanilin
Formel: C₆H₄BrClN₂O₂
Molecular Weight: 251,4700 g/mol
MDL: MFCD08275709
CAS-Nummer: 827-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR