2-Chlor-5-methoxyphenol

Lieferant: COMBI-BLOCKS

OT-0450-5G OT-0450-1G
COBBOT-0450-5GEA 285 EUR
COBBOT-0450-5G COBBOT-0450-1G
2-Chlor-5-methoxyphenol
2-Chlor-5-methoxyphenol
Formel: C₇H₇ClO₂
MDL: MFCD00086345
CAS-Nummer: 18113-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR