4-Fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluorophenylboronic acid Pinacol ester, 1-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

PN-2291-5G PN-2291-25G PN-2291-1G
COBBPN-2291-5GEA 102 EUR
COBBPN-2291-5G COBBPN-2291-25G COBBPN-2291-1G
4-Fluorphenylboronsäurepinakolester
4-Fluorphenylboronsäurepinakolester
Formel: C₁₂H₁₆BFO₂
Molecular Weight: 222,0637 g/mol
MDL: MFCD03453665
CAS-Nummer: 214360-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR