4-Fluorphenylboronsäurepinakolester
Lieferant: CHEMPUR
Synonyme:
2-(4-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluorophenylboronic acid Pinacol ester, 1-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
FL10832.20
010832-25G
010832-1G
010832-5G
CHMPFL10832.20EA
0
EUR
CHMPFL10832.20
FLUO010832-25G
FLUO010832-1G
FLUO010832-5G
4-Fluorphenylboronsäurepinakolester
4-Fluorphenylboronsäurepinakolester
Formel:
C₁₂H₁₆BFO₂ Molecular Weight: 222,0637 g/mol |
MDL:
MFCD03453665 CAS-Nummer: 214360-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...