4-Phenoxyphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

H52950.03 H52950.06
H52950.03EA 38.6 EUR
H52950.03 H52950.06
4-Phenoxyphenylboronsäure ≥98%
4-Phenoxyphenylboronsäure

Purity: 98% CAS No.: 51067-38-0 Molecular Formula: C12H11BO3 Formula Weight: 214.03 Melting Point: 141-145° MDL No.: MFCD00093312

Formel: C₁₂H₁₁BO₃
Molecular Weight: 214,03 g/mol
Schmelzpunkt: 141…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00093312
CAS-Nummer: 51067-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR