4-Phenoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

BB-2383-5G BB-2383-25G BB-2383-100G
COBBBB-2383-5GEA 33.4 EUR
COBBBB-2383-5G COBBBB-2383-25G COBBBB-2383-100G
4-Phenoxyphenylboronsäure
4-Phenoxyphenylboronsäure
Formel: C₁₂H₁₁BO₃
Molecular Weight: 214,03 g/mol
MDL: MFCD00093312
CAS-Nummer: 51067-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR