2-Chlor-4-ethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-4-ethoxybenzeneboronic acid

BB-2711-1G BB-2711-5G
COBBBB-2711-1GEA 127 EUR
COBBBB-2711-1G COBBBB-2711-5G
2-Chlor-4-ethoxyphenylboronsäure
2-Chlor-4-ethoxyphenylboronsäure
Formel: C₈H₁₀BClO₃
Molecular Weight: 200,43 g/mol
MDL: MFCD02684318
CAS-Nummer: 313545-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR