2-Chlor-5-ethoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Chloro-5-ethoxyphenyl)boronic acid
BB-2710-5G
BB-2710-1G
COBBBB-2710-5GEA
612
EUR
COBBBB-2710-5G
COBBBB-2710-1G
2-Chlor-5-ethoxyphenylboronsäure
2-Chlor-5-ethoxyphenylboronsäure
Formel:
C₈H₁₀BClO₃ Molecular Weight: 200,427 g/mol |
MDL:
MFCD08235081 CAS-Nummer: 913835-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...