Hexanophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

H12455-25G H12455-5G
SIALH12455-25GEA 73.6 EUR
SIALH12455-25G SIALH12455-5G
Hexanophenon, Sigma-Aldrich®
Hexanophenon
Formel: C₁₂H₁₆O
MDL: MFCD00009512
CAS-Nummer: 942-92-7
EINECS: 213-394-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR