2'-Fluor-5'-nitroacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(2-Fluoro-5-nitrophenyl)ethanone

RH00246.250 RH00246.1
CHMPRH00246.250EA 0 EUR
CHMPRH00246.250 MAYBRH00246.1
2'-Fluor-5'-nitroacetophenon
2'-Fluor-5'-nitroacetophenon
Formel: C₈H₆FNO₃
Molecular Weight: 183.1400 g/mol
MDL: MFCD00277867
CAS-Nummer: 79110-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR