Benzamidoxim

Lieferant: BIONET RESEARCH

Synonyme: N'-Hydroxybenzimidamide, Benzamide oxime

11T-0283.500 11T-0283.1 11T-0283.5 11T-0283.10
BIOT11T-0283.500EA 0 EUR
BIOT11T-0283.500 BIOT11T-0283.1 BIOT11T-0283.5 BIOT11T-0283.10
Benzamidoxim
Benzamidoxim
Formel: C₇H₈N₂O
Molecular Weight: 136.15 g/mol
MDL: MFCD00031485
CAS-Nummer: 613-92-3
EINECS: 210-361-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR