2-tert-Butyltoluol ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
H33036.03
H33036.06
H33036.03EA
209
EUR
H33036.03
H33036.06
2-tert-Butyltoluol ≥99%
2-tert-Butyltoluol
Purity: 99%
CAS No.: 1074-92-6
Molecular Formula: C11H16
Formula Weight: 148.25
Refractive Index: 1.5075
Formel:
C₁₁H₁₆ Molecular Weight: 148,25 g/mol Dichte: 0,89 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059209 CAS-Nummer: 1074-92-6 UN: 2667 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...