6-Chlorchinolin-2(1H)-on

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-2-hydroxyquinoline

PY-7955-1G PY-7955-5G
COBBPY-7955-1GEA 124 EUR
COBBPY-7955-1G COBBPY-7955-5G
6-Chlorchinolin-2(1H)-on
6-Chlorchinolin-2(1H)-on
Formel: C₉H₆ClNO
Molecular Weight: 179.6030 g/mol
MDL: MFCD02930633
CAS-Nummer: 1810-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR