Bis(pentafluorphenyl)dimethylsilan

Lieferant: CHEMPUR SILANE

SILS02050.1 SILS02050.5
SILNSILS02050.1EA 0 EUR
SILNSILS02050.1 SILNSILS02050.5
Bis(pentafluorphenyl)dimethylsilan
Bis(pentafluorphenyl)dimethylsilan
Formel: C₁₄H₆F₁₀Si
MDL: MFCD00042066
CAS-Nummer: 10536-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR