2,6-Bis(p-tolyl)pyridin ≥97%
Lieferant: Thermo Scientific
Synonyme:
2,6-Di-p-tolylpyridine
B24894.03
B24894.06
B24894.03EA
29.6
EUR
B24894.03
B24894.06
2,6-Bis(p-tolyl)pyridin ≥97%
2,6-Bis(p-tolyl)pyridin
Purity: 97%
CAS No.: 14435-88-2
Molecular Formula: C19H17N
Formula Weight: 259.35
Melting Point: 164-166°
MDL No.: MFCD00006282
Formel:
C₁₉H₁₇ Molecular Weight: 259,35 g/mol Schmelzpunkt: 164…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006282 CAS-Nummer: 14435-88-2 EINECS: 238-409-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...