N-(4-Methyl-2-pyridyl)thioharnstoff ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(4-Methylpyridin-2-yl)thiourea

H33648.03 H33648.06
H33648.03EA 57.1 EUR
H33648.03 H33648.06
N-(4-Methyl-2-pyridyl)thioharnstoff ≥97%
N-(4-Methyl-2-pyridyl)thioharnstoff

Purity: 97% CAS No.: 21242-21-7 Molecular Formula: C7H9N3S Formula Weight: 167.23 Melting Point: 216-220° MDL No.: MFCD04117348

Formel: C₇H₉N₃S
Molecular Weight: 167.23 g/mol
Schmelzpunkt: 216…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04117348
CAS-Nummer: 21242-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR