2-Methoxy-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-5-methylbenzeneboronic acid

BB-2158-5G BB-2158-25G BB-2158-1G
COBBBB-2158-5GEA 96.3 EUR
COBBBB-2158-5G COBBBB-2158-25G COBBBB-2158-1G
2-Methoxy-5-methylphenylboronsäure
2-Methoxy-5-methylphenylboronsäure
Formel: C₈H₁₁BO₃
Molecular Weight: 165.99 g/mol
MDL: MFCD02179465
CAS-Nummer: 127972-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR