4-Propoxy-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8089-5G BB-8089-1G
COBBBB-8089-5GEA 451 EUR
COBBBB-8089-5G COBBBB-8089-1G
4-Propoxy-2-methylphenylboronsäure
4-Propoxy-2-methylphenylboronsäure
Formel: C₁₀H₁₅BO₃
MDL: MFCD06411291
CAS-Nummer: 956894-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR