3-Methoxy-4-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methoxy-4-methylbenzeneboronic acid

BB-3959-25G BB-3959-5G BB-3959-1G
COBBBB-3959-25GEA 745 EUR
COBBBB-3959-25G COBBBB-3959-5G COBBBB-3959-1G
3-Methoxy-4-methylphenylboronsäure
3-Methoxy-4-methylphenylboronsäure
Formel: C₈H₁₁BO₃
Molecular Weight: 165.99 g/mol
MDL: MFCD08274479
CAS-Nummer: 917757-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR