(Iodmethyl)cyclopropan 95%

Lieferant: Thermo Scientific
Achtung

H35309.MD H35309.03
H35309.MDEA 48.3 EUR
H35309.MD H35309.03
(Iodmethyl)cyclopropan 95%
(Iodomethyl)cyclopropane
Formel: C₄H₇I
Molecular Weight: 182,00 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410734
CAS-Nummer: 33574-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR