4-(Benzyloxy)-3-ethoxybenzaldehyd

Lieferant: FLUOROCHEM

Synonyme: 4-Benzyloxy-3-ethoxybenzaldehyde

018222-10G 018222-25G
FLUO018222-10GEA 167 EUR
FLUO018222-10G FLUO018222-25G
4-(Benzyloxy)-3-ethoxybenzaldehyd
4-(Benzyloxy)-3-ethoxybenzaldehyd
Formel: C₁₆H₁₆O₃
Molecular Weight: 256,3000 g/mol
MDL: MFCD00615145
CAS-Nummer: 60186-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR