4-(Trifluormethyl)nicotinsäure
Lieferant: FLUOROCHEM
007629-1G
007629-5G
FLUO007629-1GEA
30.3
EUR
FLUO007629-1G
FLUO007629-5G
4-(Trifluormethyl)nicotinsäure
4-(Trifluormethyl)nicotinsäure
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191,11 g/mol |
MDL:
MFCD00082626 CAS-Nummer: 158063-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...