7-Brom-3,4-dihydro-2H-1,5-benzodioxepin 96%

Lieferant: Thermo Fisher Scientific
Achtung

H27594.06 H27594.03
H27594.06EA 104 EUR
H27594.06 H27594.03
7-Brom-3,4-dihydro-2H-1,5-benzodioxepin 96%
7-Brom-3,4-dihydro-2H-1,5-benzodioxepin

Purity: 96% CAS No.: 147644-11-9 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Flash Point: 120°(248°F) Refractive Index: 1.5790 Boiling Point: 145-148°/16mm MDL No.: MFCD00508291

Formel: C₉H₉BrO₂
Siedepunkt: 145…148 °C (16 mmHg)
Flammpunkt: >120 °C (248 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00508291
CAS-Nummer: 147644-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR