(Methoxycarbonylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Methoxy-2-oxoethyl)triphenylphosphonium bromide

M-Clarity™ Qualitätsstufe = MQ100

259063-250G 259063-50G
SIAL259063-250GEA 284 EUR
SIAL259063-250G SIAL259063-50G
(Methoxycarbonylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(Methoxycarbonylmethyl)triphenylphosphoniumbromid
Formel: C₂₁H₂₀BrO₂P
Molecular Weight: 415,28 g/mol
MDL: MFCD00011801
CAS-Nummer: 1779-58-4
EINECS: 217-222-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR