(2-Butenyl)triphenylphosphoniumchlorid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
L00502.06
L00502.14
L00502.06EA
40.3
EUR
L00502.06
L00502.14
(2-Butenyl)triphenylphosphoniumchlorid ≥97%
(2-Butenyl)triphenylphosphonium chloride
Purity: 97%
CAS No.: 13138-25-5
Molecular Formula: CH3CH=CHCH2P(C6H5)3Cl
Formula Weight: 352.84
Melting Point: ca 225-230° dec.
Beilstein Reference: 4170527
Formel:
C₂₂H₂₂ClP Molecular Weight: 352,84 g/mol Schmelzpunkt: ∼ 225…230 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051868 CAS-Nummer: 13138-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...