4-(2-Pyridyl)benzaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde
H55498.03
H55498.06
H55498.03EA
44.7
EUR
H55498.03
H55498.06
4-(2-Pyridyl)benzaldehyd ≥98%
4-(2-Pyridyl)benzaldehyd
Purity: 98%
CAS No.: 127406-56-8
Molecular Formula: C12H9NO
Formula Weight: 183.21
Flash Point: 113°(235°F)
Melting Point: 50-52°
MDL No.: MFCD01863537
Formel:
C₁₂H₉NO Molecular Weight: 183,21 g/mol Schmelzpunkt: 50…52 °C Flammpunkt: 113 °C (235 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863537 CAS-Nummer: 127406-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...