4-(4-Pyridyl)benzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Pyridin-4-yl)benzaldehyde

H55462.03 H55462.06
H55462.03EA 125 EUR
H55462.03 H55462.06
4-(4-Pyridyl)benzaldehyd ≥97%
4-(4-Pyridyl)benzaldehyd

Purity: 97% CAS No.: 99163-12-9 Molecular Formula: C12H9NO Formula Weight: 183.21 Melting Point: 85-89° MDL No.: MFCD02684110

Formel: C₁₂H₉NO
Molecular Weight: 183,21 g/mol
Schmelzpunkt: 85…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02684110
CAS-Nummer: 99163-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR