4-(4-Pyridyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Pyridin-4-yl)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

645982-1G 645982-5G
SIAL645982-1GEA 84.4 EUR
SIAL645982-1G SIAL645982-5G
4-(4-Pyridyl)benzaldehyd, Sigma-Aldrich®
4-(4-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
Molecular Weight: 183,21 g/mol
MDL: MFCD02684110
CAS-Nummer: 99163-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR