4'-(4-Methyl-1-piperazinyl)acetophenon

Lieferant: BIONET RESEARCH

7B-061.500 7B-061.1 7B-061.5 7B-061.10
BIOT7B-061.500EA 0 EUR
BIOT7B-061.500 BIOT7B-061.1 BIOT7B-061.5 BIOT7B-061.10
4'-(4-Methyl-1-piperazinyl)acetophenon
4'-(4-Methyl-1-piperazinyl)acetophenon
Formel: C₁₃H₁₈N₂O
MDL: MFCD00023297
CAS-Nummer: 26586-55-0
EINECS: 247-827-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR