4-Brom-N-phenylanilin, Sigma-Aldrich®

Lieferant: Merck

657158-1G 657158-5G
SIAL657158-1GEA 26 EUR
SIAL657158-1G SIAL657158-5G
4-Brom-N-phenylanilin, Sigma-Aldrich®
4-Brom-N-phenylanilin
Formel: C₁₂H₁₀BrN
Molecular Weight: 248,1200 g/mol
MDL: MFCD07779504
CAS-Nummer: 54446-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR