2,3-Dichlormaleinsäureanhydrid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
D65003-100G
D65003-25G
D65003-5G
SIALD65003-100GEA
988
EUR
SIALD65003-100G
SIALD65003-25G
SIALD65003-5G
2,3-Dichlormaleinsäureanhydrid, Sigma-Aldrich®
2,3-Dichlormaleinsäureanhydrid
Formel:
C₄Cl₂O₃ Molecular Weight: 166,9500 g/mol |
MDL:
MFCD00005520 CAS-Nummer: 1122-17-4 EINECS: 214-343-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...