4'-(2-Chlorethyl)acetophenon

Lieferant: BIONET RESEARCH

Synonyme: 1-[4-(2-Chloroethyl)phenyl]ethan-1-one

1D-150.500 1D-150.1 1D-150.5 1D-150.10
BIOT1D-150.500EA 0 EUR
BIOT1D-150.500 BIOT1D-150.1 BIOT1D-150.5 BIOT1D-150.10
4'-(2-Chlorethyl)acetophenon
4'-(2-Chlorethyl)acetophenon
Formel: C₁₀H₁₁ClO
Molecular Weight: 182,65 g/mol
MDL: MFCD00016350
CAS-Nummer: 69614-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR