4-(4-Brombenzyloxy)benzaldehyd

Lieferant: BIONET RESEARCH

Synonyme: 4-((4-Bromobenzyl)oxy)benzaldehyde

12P-045.500 12P-045.1 12P-045.5 12P-045.10
BIOT12P-045.500EA 0 EUR
BIOT12P-045.500 BIOT12P-045.1 BIOT12P-045.5 BIOT12P-045.10
4-(4-Brombenzyloxy)benzaldehyd
4-(4-Brombenzyloxy)benzaldehyd
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291.1399 g/mol
MDL: MFCD02091001
CAS-Nummer: 149833-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR