3-(Benzyloxy)-5-chlorbenzaldehyd
Lieferant: COMBI-BLOCKS
CA-5003-5G
CA-5003-1G
COBBCA-5003-5GEA
595
EUR
COBBCA-5003-5G
COBBCA-5003-1G
3-(Benzyloxy)-5-chlorbenzaldehyd
3-(Benzyloxy)-5-chlorbenzaldehyd
Formel:
C₁₄H₁₁ClO₂ |
MDL:
MFCD16618949 CAS-Nummer: 1228956-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...